Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Diet Soda Spiked with Neotame on Ascentis® Express RP-Amide
HPLC Analysis of Diet Soda Spiked with Neotame on Ascentis® Express RP-Amide
Conditions
column Ascentis Express RP-Amide, 3 cm x 4.6 mm I.D., 2.7 µm particles (53921-U)
column temp. 40 C
mobile phase [A] 100 mM ammonium acetate, pH 5.6 with acetic acid; [B] water; [C] acetonitrile
gradient 20% A constant; 75 to 20% B, 5 to 60% C in 1 min; held at 20% B, 60% C for 0.1 min
flow rate 3 ml/min
pressure 3336 psi (230 bar)
sample diet soda spiked with neotame 350 µg/mL
injection 1 ml/min
detector UV, 214 nm
Description
Analysis Note This application demonstrates the suitability of Ascentis Express RP-Amide for the efficient separation of common drink additives used in diet cola.
Categories Analytical Chromatography, Flavors, Fragrances
Featured Industry Food and Beverages