HPLC Analysis of Diquat on SeQuant ZIC-HILIC, Effect of Mobile Phase Ionic Strength on Retention
HPLC Analysis of Diquat on SeQuant ZIC-HILIC, Effect of Mobile Phase Ionic Strength on Retention
Conditions
column SeQuant ZIC-HILIC, PEEK 150, x 4.6 mm, 5 µm, 200 Ã… (1.50455.0001)
column temp. 40 C
mobile phase [A] acetonitrile; [B] 62.5 to 250 mM ammonium acetate, pH 3.7; (40:60, A:B)
flow rate 1.0 ml/min
injection 5 µl
detector 313 nm
Description
Featured Industry Agriculture