HPLC Analysis of DNA on Proteomix® WAX-NP5
HPLC Analysis of DNA on Proteomix® WAX-NP5
Conditions
column Proteomix WAX-NP5, 15 cm x 4.6 mm I.D., 5 µm particles (Z777262)
mobile phase (A) 20 mmol Tris, pH 6.0; (B) A + 1.0 M NaCl
gradient 20 to 100% B in 40 min
flow rate 0.6 mL/min
pressure 1015 psi (70 bar)
temp. ambient
sample DNA
injection 5 µL
detector UV, 260 nm
Description
Categories Analytical Chromatography, Nucleic Acids
Featured Industry Life Science and Biopharma
Legal Information Proteomix is a registered trademark of Sepax Technologies