HPLC Analysis of Dopa and Carbidopa Enantiomers on Astec® CHIROBIOTIC® T2
HPLC Analysis of Dopa and Carbidopa Enantiomers on Astec® CHIROBIOTIC® T2
Conditions
column CHIROBIOTIC T2, 25 cm x 4.6 mm I.D., 5 µm particles (16024AST)
column temp. 25 °C
mobile phase [A] 20 mM ammonium acetate, pH 4; [B] methanol; (10:90, A:B)
flow rate 1 mL/min
sample 1 mg/mL in methanol
injection 5 µL
detector UV, 230 nm
Description
Categories Analytical Chromatography, Chiral Applications, Clinical, Pharmaceutical (small molecule), antiparkinsonian, Antiparkinsonians, Clinical, Forensic & Toxicology, Antiparkinson Drugs, All Pharmaceutical Applications, Therapeutic Drug Monitoring
Featured Industry Clinical
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC