Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Dorzolamide Enantiomers on Astec® CHIROBIOTIC® V2
HPLC Analysis of Dorzolamide Enantiomers on Astec® CHIROBIOTIC® V2
Conditions
column CHIROBIOTIC V2, 25 cm x 4.6 mm I.D., 5 µm particles (15024AST)
column temp. 22 C
mobile phase [A] 3.81 mM ammonium trifluoroacetic acid; [B] methanol; (5:95, A:B)
flow rate 0.3 ml/min
sample 1 mg/mL in methanol
injection 10 µl
detector UV, 254 nm
Description
Categories Analytical Chromatography, Chiral Applications, Antiglaucoma Drugs, Pharmaceutical (small molecule), All Pharmaceutical Applications
Featured Industry Pharmaceutical (small molecule)
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC