HPLC Analysis of Drugs of Abuse on Ascentis® Si (HILIC Mode)
HPLC Analysis of Drugs of Abuse on Ascentis® Si (HILIC Mode)
Conditions
column Ascentis Si, 5 cm x 4.6 mm I.D., 5 µm particles (581511-U)
column temp. 35 °C
mobile phase [A] 0.1% ammonium acetate in water (pH unadjusted): [B] 0.1% ammonium acetate in acetontrile; (10:90, A:B)
flow rate 1 mL/min
sample as indicated in 0.1% ammonium acetate in 10:90:water:acetonitrile
injection 10 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Forensics and Toxicology, drugs of abuse, Analgesics
Featured Industry Forensics and Toxicology