HPLC Analysis of E. coli Lysate on Proteomix® SAX Columns, Particle Size Comparison
HPLC Analysis of E. coli Lysate on Proteomix® SAX Columns, Particle Size Comparison
Conditions
column Proteomix SAX-NP5, 5 cm x 4.6mm I.D., 5 µm particles (Z777227)
column Proteomix SAX-NP10, 5 cm x 4.6 mm I.D., 10 µm particles (Z777203)
column temp. 25 °C
mobile phase (A) 20 mM Tris, pH 9.0; (B) A + 0.5 M NaCl
gradient 0 - 100%B in 30 min
flow rate 0.5 mL/min
sample 2.5 mg/mL
injection 10 µL
detector UV, 280 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Pharmaceutical (small molecule)
Legal Information Proteomix is a registered trademark of Sepax Technologies