HPLC Analysis of E. coli lysate with various pore size Zenix® columns
HPLC Analysis of E. coli lysate with various pore size Zenix® columns
Conditions
column Zenix SEC-150, 30 cm x 7.8 mm I.D., 3 µm particles (Z777018)
column Zenix SEC-300, 30 cm x 7.8 mm I.D., 3 µm particles (Z777033)
column temp. Ambient
mobile phase 150 mM Phosphate Buffer, pH 7
flow rate 1.0 ml/min
sample E. coli lysate, 2.5 mg/mL
injection 10 µl
detector UV, 214nm
Description
Analysis Note An cell lysate, produced by destroying the membranes of E. Coli cells, is analyzed on two Zenix gel filtration columns packed with 3 micron particles. The top chromatogram shows the peak profile on a 150 Ã… pore size Zenix SEC-150 gel filtration column, while the bottom chromatogram shows the same sample on a 300 Ã… pore size Zenix SEC-300 column. As expected, the peaks shift to longer retention times on the SEC-300 column. Although roughly the same number of components are visible in each chromatogram, the SEC-150 column does a better job of pulling the peaks apart, which helps when fractions need to be collected for further analysis.
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information Zenix is a registered trademark of Sepax Technologies