HPLC Analysis of Eflornithine (DFMO) on SeQuant ZIC-HILIC
HPLC Analysis of Eflornithine (DFMO) on SeQuant ZIC-HILIC
Conditions
column SeQuant ZIC-HILIC, PEEK, 50 x 4.6 mm, 5 µm, 200 Ã… (1.50451.0001)
mobile phase [A] acetonitrile; [B] ammonium acetate; (75:25, A,B)
flow rate 0.5 ml/min
injection 5 µl
detector 210 nm
Description
Categories Analytical Chromatography, Drugs, miscellaneous