HPLC Analysis of EPA 8330 Mix A and EPA 8330 Mix B on Ascentis® Express C18
HPLC Analysis of EPA 8330 Mix A and EPA 8330 Mix B on Ascentis® Express C18
Conditions
column Ascentis Express C18, 15 cm x 4.6 mm I.D., 5 µm particles (50537-U)
column temp. 42 C
mobile phase mobile phase: [A] water [B] methanol (72:28, A:B)
flow rate 2.00 ml/min
pressure 3277 psi (226 bar)
sample 0.033 mg/mL in water/acetonitrile; 33:67
injection 10 µl
detector UV, 254 nm
Description
Categories Analytical Chromatography, Explosives
Featured Industry Environmental