HPLC Analysis of Ephedrine Enantiomers on CHIRALPAK® AGP
HPLC Analysis of Ephedrine Enantiomers on CHIRALPAK® AGP
Conditions
column CHIRAL-AGP, 10 cm x 4 mm I.D., 5 µm particles (58170AST)
column temp. 25 °C
mobile phase 1 mM octanoic acid in 10 mM (NH4)2HPO4, pH 6.9
flow rate 0.9 mL/min
sample 1 mg/mL in methanol
injection 10 µL
detector UV, 225 nm
Description
Categories Analytical Chromatography, Antihistamines
Featured Industry Pharmaceutical (small molecule)
Legal Information CHIRALPAK is a registered trademark of Daicel Corp.