Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Epinephrine, Epinastine, and Benzalkonium Chlorides on Ascentis® Express HILIC
HPLC Analysis of Epinephrine, Epinastine, and Benzalkonium Chlorides on Ascentis® Express HILIC
Conditions
column Ascentis Express HILIC, 15 cm x 4.6 mm I.D., 2.7 µm particles (53981-U)
column temp. 35 C
mobile phase [A] 50 mM ammonium acetate; [B] acetonitrile; (10:90, A:B)
flow rate 1 ml/min
sample 10 mg/L ea. in acetonitrile
injection 10 µl
detector ESI(+), XIC m/z 304 & 332 (BAC), 250 (epinastine), 180 (epinephrine)
Description
Categories Analytical Chromatography, Antihistamines, Hormones, Neurotransmitters, Surfactants