Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Epoxy resin, Epikote® 1001 on TSKgel® G3000HXL
HPLC Analysis of Epoxy resin, Epikote® 1001 on TSKgel® G3000HXL
Conditions
column TSKgel G3000HXL, 30 cm x 7.8 mm I.D., 6 µm particles (816136)
mobile phase tetrahydrofuran
flow rate 1 ml/min
detector UV, 254 nm
Description
Categories Analytical Chromatography, Volatiles
Featured Industry Chemicals and Industrial Polymers
Legal Information Epikote is a registered trademark of Hexion, Inc., TSKgel is a registered trademark of Tosoh Corporation