HPLC Analysis of Erectile Dysfunction Drugs on Discovery® C18
HPLC Analysis of Erectile Dysfunction Drugs on Discovery® C18
Conditions
column Discovery C18, 15 cm x 4.6 mm I.D., 5 µm particles (504955)
column temp. 35 °C
mobile phase [A] water (10 mM NH4COOH); [B] acetonitrile; (50:50, A:B)
flow rate 1.0 mL/min
sample 50 µg/mL each in 50:50 water:acetonitrile
injection 5 µL
detector UV, 230 nm
Description
Categories Analytical Chromatography, ED Drugs
Featured Industry Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC