HPLC Analysis of Fat Soluble Vitamins by Normal Phase Chromatography on Ascentis® Si, 15 cm Column
HPLC Analysis of Fat Soluble Vitamins by Normal Phase Chromatography on Ascentis® Si, 15 cm Column
Conditions
column Ascentis Si, 15 cm x 4.6 mm I.D., 5 µm particles (581512-U)
column temp. 30 °C
mobile phase [A] hexane, [B] ethyl acetate
gradient 10 to 30% B in 10 min; held for 2 min; to 10% B in 1 min
flow rate 1.0 mL/min
sample as indicated in 96:4 (hexane:isopropanol)
injection 10 µL
detector UV, 290 nm
Description
Categories Analytical Chromatography, Vitamins and Natural products
Featured Industry Vitamins, Nutraceuticals, and Natural Products