HPLC Analysis of Fat Soluble Vitamins on Ascentis® Express RP-Amide, Acetonitrile Mobile Phase
HPLC Analysis of Fat Soluble Vitamins on Ascentis® Express RP-Amide, Acetonitrile Mobile Phase
Conditions
column Ascentis Express RP-Amide, 15 cm x 4.6 mm I.D., 2.7 µm particles (53931-U)
column temp. 30 C
mobile phase acetonitrile
flow rate 1.5 ml/min
pressure 1682 psi (116 bar)
sample 0.5 mg/mL tocopherol acetate, alpha-tocopherol, delta-tocopherol and K1, 0.05 mg/mL D2, D3 and retinol acetate, 1.5 mg/mL retinol palmitate, all in ethanol
injection 5 µl
detector UV, 280 nm
Description
Analysis Note Analysis was conducted using standard solution, detection via UV.
Categories Analytical Chromatography, Vitamins and Natural products
Featured Industry Vitamins, Nutraceuticals, and Natural Products