HPLC Analysis of Fat-Soluble Vitamins on Ascentis® Express C18, Methanol Mobile Phase
HPLC Analysis of Fat-Soluble Vitamins on Ascentis® Express C18, Methanol Mobile Phase
Conditions
column Ascentis Express C18, 15 cm x 4.6 mm I.D., 2.7 µm particles (53829-U)
column temp. 30 C
mobile phase methanol
flow rate 1.5 ml/min
pressure 3249 psi (224 bar)
sample 0.5 mg/mL tocopherol acetate, alpha-tocopherol, delta-tocopherol and K1, 0.05 mg/mL D2, D3 and retinol acetate, 1.5 mg/mL retinol palmitate, all in ethanol.
injection 5 µl
detector UV, 280 nm
Description
Analysis Note Analysis was conducted using standard solution, detection via UV.
Categories Analytical Chromatography, Vitamins and Natural products
Featured Industry Vitamins, Nutraceuticals, and Natural Products