HPLC Analysis of Fatty Acid Methyl Esters (FAMES) on SUPELCOSIL™ LC-18
HPLC Analysis of Fatty Acid Methyl Esters (FAMES) on SUPELCOSIL™ LC-18
Conditions
column SUPELCOSIL LC-18, 25 cm x 4.6 mm I.D., 5 µm particles (58298)
mobile phase [A] acetonitrile; [B] acetone; (59.0:41.0, v/v, A:B)
flow rate 1 mL/min
injection 10 µL of 9% C8 to C18 saturated and unsaturated FAMEs in mobile phase
detector RI
Description
Categories Analytical Chromatography, Fatty Acids, FAMEs
Featured Industry Chemicals and Industrial Polymers
Food and Beverages
Legal Information SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC