HPLC Analysis of Fentanyl Tranquilizer on Ascentis® C18
HPLC Analysis of Fentanyl Tranquilizer on Ascentis® C18
Conditions
column Ascentis C18, 15 cm x 4.6 mm I.D., 5 µm particles (581324-U)
column temp. 35 °C
mobile phase [A] 10 mM monobasic potassium phosphate (pH 3.0 with phosphoric acid): [B] acetonitrile; (67:33, A:B)
flow rate 1 mL/min
sample 20 µg/mL in mobile phase
injection 10 µL
detector UV, 215 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Analgesics