HPLC Analysis of Fipronil Enantiomers on Astec® (R,R) P-CAP™
HPLC Analysis of Fipronil Enantiomers on Astec® (R,R) P-CAP™
Conditions
column Astec (R,R) P-CAP, 25 cm x 4.6 mm I.D., 5 µm particles (31024AST)
column temp. 25 °C
mobile phase [A] heptane; [B] ethanol; (80:20, A:B)
flow rate 1 mL/min
sample 1 mg/mL in methanol
injection 1 µL
detector UV, 235 nm
Description
Categories Analytical Chromatography, Chiral Applications, Pesticides & Herbicides, Environmental, Pesticides, All Environmental Applications
Featured Industry Chemicals and Industrial Polymers
Legal Information P-CAP is a trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC