HPLC Analysis of Flakka (α-PVP) and α-Pyrrolidinobutiophenone on Ascentis® Express HILIC 5 μm
HPLC Analysis of Flakka (α-PVP) and α-Pyrrolidinobutiophenone on Ascentis® Express HILIC 5 μm
Conditions
column Ascentis Express HILIC, 5 cm x 3.0 mm I.D., 5 μm particles (50262-U)
mobile phase 5 mM ammonium formate in 95:5, acetonitrile:water
flow rate 0.8 mL/min
pressure 453 psi (31.2 bar)
column temp. 35 °C
detector UV, 250 nm
injection: 2 μL
sample 50 μg/mL in acetonitrile
Description
Featured Industry Forensics and Toxicology
Legal Information Ascentis is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for HPLC
Analysis Note The rapid separation of the street drug Flakka (α-PVP) and related compound is shown here on an Ascentis Express HILIC column. Highest grade UHPLC solvents were used to supply low background interference and low particulate contamination for robust, trouble-free operation. Cerilliant CRMs provided reliable identification and quantification.