HPLC Analysis of Flavonoids in Orange Juice on Ascentis® C18
HPLC Analysis of Flavonoids in Orange Juice on Ascentis® C18
Conditions
column Ascentis C18, 25 cm x 4.6 mm I.D., 5 µm particles (581325-U)
column temp. 25 °C
mobile phase [A] formic acid: [B] 2-propanol: [C] acetonitrile: [D] water (0.1:10:12:78, A:B:C:D:)
flow rate 0.8 mL/min
sample orange juice, passed through a 0.45 µm filter
injection 10 µL
detector PDA, 283 nm
Description
Categories Analytical Chromatography, Food & Beverage Analysis, Flavors, Fragrances, Beverage Testing, Fruit Juice
Featured Industry Food and Beverages