HPLC Analysis of Fluorinated Pyrimidine Nucleosides on Ascentis® RP-Amide
HPLC Analysis of Fluorinated Pyrimidine Nucleosides on Ascentis® RP-Amide
Conditions
column Ascentis RP-Amide, 5 cm x 4.6 mm I.D., 5 µm particles (565323-U)
column temp. 30 °C
mobile phase [A] water with 0.1% ammonium formate (pH 3.04 with formic acid), [B] acetonitrile
flow rate 1.0 mL/min
sample as indicated in mobile phase A
injection 5 µL
detector UV, 260 nm
Description
Categories Analytical Chromatography, Clinical, nucleic acids, Nucleic Acids
Featured Industry Clinical
Life Science and Biopharma