HPLC Analysis of Fluoroquinolone Antibiotics on Ascentis® Phenyl
HPLC Analysis of Fluoroquinolone Antibiotics on Ascentis® Phenyl
Conditions
column Ascentis Phenyl, 15 cm x 4.6 mm I.D., 5 µm particles (581616-U)
column temp. 35 °C
mobile phase [A] 13 mM ammonium formate (pH 3.0 with concentrated formic acid): [B] acetonitrile; (80:20, A:B)
flow rate 1 mL/min, split to the MS
sample as indicated in 80:20, 13 mM ammonium formate (pH 3.0 with concentrated formic acid):acetonitrile
injection 5 µL
detector MS, ESI (+) in selected ion recording (SIR) mode
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Antibiotics, Antibiotics, Antimicrobials
Featured Industry Food and Beverages
Pharmaceutical (small molecule)