Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Fluoroquinolone Antibiotics on Ascentis® Express C18
HPLC Analysis of Fluoroquinolone Antibiotics on Ascentis® Express C18
Conditions
column Ascentis Express C18, 10 cm x 3.0 mm I.D., 2.7 µm particles (53814-U)
column temp. 35 C
mobile phase [A] 0.1% formic acid in water; [B] 0.1% formic acid in acetonitrile; (85:15, A:B)
flow rate 1.2 ml/min
sample 200 ng/mL in 25:75, water: acetonitrile
injection 4 µl
detector UV, 254 nm
Description
Analysis Note This application demonstrates the suitability of the Ascentis Express C18 for the analysis of fluoroquinolones.
Categories Analytical Chromatography, Antibiotics, Antimicrobials
Featured Industry Food and Beverages
Pharmaceutical (small molecule)