HPLC Analysis of Fluoxetine (Prozac®) and Norfluoxetine on Discovery® Zr-PS
HPLC Analysis of Fluoxetine (Prozac®) and Norfluoxetine on Discovery® Zr-PS
Conditions
column Discovery Zr-PS, 7.5 cm x 4.6 mm I.D., 3 µm particles (65741-U)
column temp. 35 C
mobile phase [A] 25 mM potassium phosphate (pH 12): [B] acetonitrile; (70:30, A:B)
flow rate 1 ml/min
injection 10 µl
detector UV, 230 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Antidepressants
Featured Industry Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC, Prozac is a registered trademark of Eli Lilly and Co.