HPLC Analysis of Fluoxetine (Prozac®) on Discovery® C18
HPLC Analysis of Fluoxetine (Prozac®) on Discovery® C18
Conditions
column Discovery C18, 15 cm x 4.6 mm I.D., 5 µm particles (504955)
column temp. 30 °C
mobile phase [A] acetonitrile; [B] 25 mM KH2PO4, pH 7.0 (40:60, A:B)
flow rate 2 mL/min
injection 1 µL
detector UV, 214 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Antidepressants
Featured Industry Pharmaceutical (small molecule)
Legal Information Prozac is a registered trademark of Eli Lilly and Co., Discovery is a registered trademark of Sigma-Aldrich Co. LLC