HPLC Analysis of FMOC-Arginine Enantiomers on Astec® CHIROBIOTIC® R
HPLC Analysis of FMOC-Arginine Enantiomers on Astec® CHIROBIOTIC® R
Conditions
column CHIROBIOTIC R, 25 cm x 4.6 mm I.D., 5 µm particles (13024AST)
column temp. 23 °C
mobile phase 0.1% ammonium trifluoroacetic acid in methanol
flow rate 1 mL/min
sample 1 mg/mL in methanol
injection 2 µL
detector UV, 260 nm
Description
Categories Analytical Chromatography, Chiral Applications, Amino Acids, Life Science & Biopharma, All Biochemical Applications, All Amino Acids, FMOC-Amino Acids
Featured Industry Life Science and Biopharma
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC