HPLC Analysis of Folinic Acid Enantiomers on Astec® CHIROBIOTIC® T
HPLC Analysis of Folinic Acid Enantiomers on Astec® CHIROBIOTIC® T
Conditions
column CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 µm particles (12024AST)
column temp. 25 °C
mobile phase [A] methanol; [B] acetic acid; [C] triethylamine; (100:1:0.5, A:B:C)
flow rate 2 mL/min
sample 5 mg/mL in methanol
injection 2 µL
detector UV, 230 nm
Description
Categories Analytical Chromatography, Chiral Applications, Food & Beverage, Pharmaceutical (small molecule), Antineoplastics, Life Science & Biopharma, Vitamins and Natural products, All Food & Beverage Applications, All Biochemical Applications, Vitamins & Coenzymes, All Pharmaceutical Applications, Vitamins
Featured Industry Vitamins, Nutraceuticals, and Natural Products
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC