HPLC Analysis of Folinic Acid Enantiomers on Astec® CHIROBIOTIC® T (RP mode)
HPLC Analysis of Folinic Acid Enantiomers on Astec® CHIROBIOTIC® T (RP mode)
Conditions
column CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 µm particles (12024AST)
column temp. 20 °C
mobile phase [A] 1% triethylamine acetate, pH 4.5; [B] ethanol; (50:50, A:B)
flow rate 1 mL/min
sample 1.25 mg/mL in methanol
injection 5 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Chiral Applications, Antineoplastics, Vitamins and Natural products, Food & Beverage, Pharmaceutical (small molecule), Life Science & Biopharma, All Food & Beverage Applications, All Biochemical Applications, Vitamins & Coenzymes, All Pharmaceutical Applications, Vitamins
Featured Industry Vitamins, Nutraceuticals, and Natural Products
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC