HPLC Analysis of Free Fatty Acids on SUPELCOSIL™ LC-8
HPLC Analysis of Free Fatty Acids on SUPELCOSIL™ LC-8
Conditions
column SUPELCOSIL LC-8, 15 cm x 4.6 mm I.D., 3 µm particles (58983)
column temp. 35 °C
mobile phase acetonitrile:tetrahydrofuran: 0.1% phosphoric acid (50.4:21.6:28)
flow rate 1 mL/min
injection 10 µL
detector UV, 215 nm
Description
Categories Analytical Chromatography, Fatty Acids, FAMEs
Featured Industry Food and Beverages
Legal Information SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC