Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Fumaric, Oxalic, and Citric Acid on SeQuant ZIC-HILIC
HPLC Analysis of Fumaric, Oxalic, and Citric Acid on SeQuant ZIC-HILIC
Conditions
column SeQuant ZIC-HILIC, PEEK, 150 x 4.6 mm, 5 µm, 200 Ã… (1.50455.0001)
mobile phase [A] acetonitrile; [B] ammonium acetate; (70:30, A,B)
flow rate 1.5 ml/min
pressure 1240 psi (8.6 MPa)
injection 20 µl
detector 220 nm
Description
Categories Analytical Chromatography, Organic Acids
Featured Industry Food and Beverages