HPLC Analysis of Furans on Ascentis® Express C18
HPLC Analysis of Furans on Ascentis® Express C18
Conditions
column Ascentis Express C18, 5 cm x 3.0 mm I.D., 2.7 µm particles (53811-U)
column temp. 35 C
mobile phase [A] water; [B] acetonitrile; (80:20, A:B)
flow rate 0.4 ml/min
sample 20 µg/mL in 80:20, water: acetonitrile
injection 1 µl
detector UV, 230 nm
Description
Analysis Note This application demonstrates the suitability of Ascentis Express C18 for the efficient separation of furans as listed in ASTM method 05837-99.
Categories Analytical Chromatography, Furans
Featured Industry Chemicals and Industrial Polymers