HPLC Analysis of Furosemide and Indapamide on Discovery® C18
HPLC Analysis of Furosemide and Indapamide on Discovery® C18
Conditions
column Discovery C18, 15 cm x 4.6 mm ID, 5 µm particles (504955)
column temp. 35 °C
mobile phase [A] 10mM monobasic potassium phosphate, (pH 3.0 with H3PO4): [B] acetonitrile; (40:60, A:B)
flow rate 1 mL/min
sample 10 µg/mL each in mobile phase
injection 10 µL
detector UV, 234 nm
Description
Categories Analytical Chromatography, Diuretics
Featured Industry Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC