HPLC Analysis of Fusion Protein on Zenix®-C SEC-300
HPLC Analysis of Fusion Protein on Zenix®-C SEC-300
Conditions
column 25 °C
column Zenix-C SEC-300, 30 cm x 7.8 mm I.D., 3 µm particles (Z777092)
mobile phase 150 mM sodium phosphate buffer, pH 7.0 + 200 mM NaCl
flow rate 1.0 mL/min
pressure 1102 psi (76 bar)
sample 1 mg/mLfusion protein
injection 10 µL
detector UV, 214 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information Zenix is a registered trademark of Sepax Technologies