HPLC Analysis of Geldanamycin and 17-(Allylamino)-17-demethoxy geldanamycin on Ascentis® Express C18
HPLC Analysis of Geldanamycin and 17-(Allylamino)-17-demethoxy geldanamycin on Ascentis® Express C18
Conditions
column Ascentis Express C18, 10cm X 3.0mm I.D, 2.7 µm particles (53814-U)
column temp. 35 °C
mobile phase [A] 0.1% formic acid in water [B] acetonitrile, 55:45 A:B
flow rate 0.8 mL/min
pressure 3300 psi (228 bar)
sample 50 ug/mL each in 10% methanol
injection 5 µL
detector UV, 254 nm
Description
Analysis Note The compound 7-(allylamino)-17-demethoxygeldanamycin (AAG) is a derivative of geldanamycin, a potent herbicidal agent isolated from the soil organism streptomyces hygroscopicus. Geldanamycin has been explored as a possible human antibiotic, but toxicity concerns have hindered its introduction. However, the AAG derivative is better tolerated and has undergone further clinical development at the time of this writing. Shown here is the rapid separation of both compounds on an Ascentis Express C18 column. High purity solvents and additives used in the mobile phase provided robust operation.
Categories Analytical Chromatography, Herbicides
Featured Industry Agriculture
Environmental
Pharmaceutical (small molecule)