HPLC Analysis of Ginsenoside Standards on Ascentis® Express C18
HPLC Analysis of Ginsenoside Standards on Ascentis® Express C18
Conditions
column Ascentis Express C18, 15 cm x 4.6 mm I.D., 2.7 µm particles (53829-U)
column temp. 60 °C
mobile phase [A]: water; [B]: acetonitrile
gradient held at 25% for 1.5 min; 25 to 85% B in 12 min; held at 85% B for 1 min
flow rate 1.0 mL/min
pressure 3481 psi (240 bar)
sample 50 µg/mL in water:methanol (82:18)
injection 25 µL
detector UV, 205 nm
Description
Analysis Note This application demonstrates the suitability of Ascentis Express C18 for the efficient separation of ginseng.
Categories Analytical Chromatography, Glycosides
Featured Industry Vitamins, Nutraceuticals, and Natural Products