Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Ginsenosides on Ascentis® Express C18
HPLC Analysis of Ginsenosides on Ascentis® Express C18
Conditions
column Ascentis Express C18, 15 cm x 4.6 mm I.D., 5 µm particles (50537-U)
column temp. 60 C
mobile phase [A] water, [B] acetonitrile
gradient 25% B held for 3 min; to 85% B in 12 min;85% B held for 1 min
flow rate 1 ml/min
pressure 1470 psi (101 bar)
sample 50 µg/mL in 82:18, water:acetonitrile
injection 5 µl
detector UV, 205 nm
Description
Categories Analytical Chromatography, Vitamins and Natural products
Featured Industry Vitamins, Nutraceuticals, and Natural Products