HPLC Analysis of Glyocoalkaloids Solanine and Cacoline on Chromolith® NH2
HPLC Analysis of Glyocoalkaloids Solanine and Cacoline on Chromolith® NH2
Conditions
column Chromolith NH2, 5 cm x 4.6 mm I.D. (1.52027.0001)
column temp. 23 C
mobile phase [A] acetonitrile; [B] phosphate buffer; pH 6 (80:20, A:B)
flow rate 2 ml/min
pressure 174 psi (12 bar)
sample solanine 0.2 mg/mL, cacoline 0.2 mg/mL, in mobile phase
injection 8 µl
detector UV, 208 nm
Description
Categories Analytical Chromatography, Carbohydrates
Featured Industry Food and Beverages
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany