HPLC Analysis of GMP, GDP and GTP on SeQuant ZIC-HILIC
HPLC Analysis of GMP, GDP and GTP on SeQuant ZIC-HILIC
Conditions
column SeQuant ZIC-HILIC, PEEK, 100 x 4.6 mm, 5 µm, 200 Ã… (1.50453.0001)
mobile phase [A] acetonitrile; [B] ammonium acetate; (70:30, A,B)
flow rate 0.7 ml/min
injection 5 µl
detector 259 nm
Description
Categories Analytical Chromatography, Nucleotides