HPLC Analysis of Hemoglobin HbA1C on Mono S® HR5/5
HPLC Analysis of Hemoglobin HbA1C on Mono S® HR5/5
Conditions
column Mono S HR5/5, 5 cm x 5 mm I.D., 10 µm (54808)
mobile phase [A] 0.01M malonate-sodium hydroxide, pH 5.7; [B] 0.3M lithium chloride in mobile phase A
flow rate 2.0 ml/min
injection 50 µl hemolysate, dilued 1:5
detector VIS, 405 nm
Description
Categories Analytical Chromatography, Peptides
Featured Industry Life Science and Biopharma
Legal Information Mono S is a registered trademark of GE Healthcare