HPLC Analysis of Herbal Supplement Containing Milk Thistle on Ascentis® Express 2.7 μm C18
HPLC Analysis of Herbal Supplement Containing Milk Thistle on Ascentis® Express 2.7 μm C18
Conditions
column Ascentis Express C18, 10 cm x 3.0 mm I.D., 2.7 µm particles (53814-U)
column temp. 35 °C
mobile phase [A] water with 0.1% formic acid; [B] methanol
gradient held at 35% B for 3 min; 35 to 45% B in 10 min; held at 45% B for 2 min; 45 to 100% B in 5 min
flow rate 0.6 mL/min
pressure 4105 psi (283 bar)
sample 20 mg/mL in water:ethanol (5:95); sonicate 15 minutes; filter 0.45 µm; dilute to water:ethanol (80:20)
injection 20 µL
detector UV, 254 nm
Description
Analysis Note This application demonstrates the suitability of Ascentis Express C18 for the efficient separation of milk thistle containing herbal supplement.
Categories Analytical Chromatography, Herbal medicines
Featured Industry Vitamins, Nutraceuticals, and Natural Products