Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Histidine Enantiomers on Astec® CHIROBIOTIC® T (15 °C)
HPLC Analysis of Histidine Enantiomers on Astec® CHIROBIOTIC® T (15 °C)
Conditions
column CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 µm particles (12024AST)
column temp. 15 C
mobile phase [A] water; [B] methanol; [C] formic acid; (30:70:0.006, A:B:C)
flow rate 0.5 ml/min
sample 200 mug /mL in 50:50 water:methanol
injection 10 µl
detector UV, 205 nm
Description
Categories Analytical Chromatography, Amino Acids
Featured Industry Life Science and Biopharma
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC