HPLC Analysis of HPLC Protein Standard on BIOshell™ A400 Protein C4
HPLC Analysis of HPLC Protein Standard on BIOshell™ A400 Protein C4
Conditions
column BIOShell A400 °C18, 10 cm x 2.1 mm I.D., 3.4 µm particles (67463-U)
column temp. 30 °C
mobile phase [A] (water, 0.1% TFA); (acetonitrile, 0.08% TFA), 75:25; [B] (water, 0.1% TFA); (acetonitrile, 0.08% TFA), 25:75
gradient 0 to 75% B in 15 min
flow rate 0.3 mL/min
pressure 2800 psi (193 bar)
sample ribonuclease A, cytochrome c, holo-transferrin and myoglobin 1 mg in mobile phase A
injection 2 µL
detector UV, 215 nm
Description
Analysis Note Analysis of protein standard of various hydrophobicity on BIOshell A400 Protein C18 to compare the selectivity and retention times.
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information BIOshell is a trademark of Sigma-Aldrich Co. LLC