Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of HSA Fusion Protein on Zenix®-C SEC-300 vs. Zenix® SEC-300
HPLC Analysis of HSA Fusion Protein on Zenix®-C SEC-300 vs. Zenix® SEC-300
Conditions
column Zenix-C SEC-300, 30 cm x 7.8 mm I.D., 3 µm particles (Z777092)
column Zenix SEC-300, 30 cm x 7.8 mm I.D., 3 µm particles (Z777033)
column temp. 25 C
mobile phase 150 mM sodium phosphate buffer, pH 7.0
flow rate 1 ml/min
sample HSA fusion peptide 5 mg/mL (MW 75 kD, pl 5.0, HSA conjugated peptide in diabetes treatment)
injection 10 µl
detector UV, 280 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information Zenix is a registered trademark of Sepax Technologies