Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of HSA Fusion Protein on Proteomix® SAX-NP5
HPLC Analysis of HSA Fusion Protein on Proteomix® SAX-NP5
Conditions
column Proteomix SAX-NP5 25 cm x 4.6mm I.D., 5 µm particles (Z777229)
column temp. 30 C
mobile phase Piperazine, Triethanolamine, bis-tris propane and N-methylpiperazine, 20 mM each,(A) pH 9.7; (B) pH 3.0 ( adjust by HCl )
flow rate 0.5 ml/min
pressure 2016 psi (139 bar)
sample HSA fusion peptide 5 mg/mL, 10 µl (MW 75 kD, pI 5.0, HSA conjugated peptide in diabetes treatment)
detector UV, 280 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Pharmaceutical (small molecule)
Legal Information Proteomix is a registered trademark of Sepax Technologies