HPLC Analysis of Hydrobenzoin Enantiomers on Astec® CYCLOBOND® I 2000
HPLC Analysis of Hydrobenzoin Enantiomers on Astec® CYCLOBOND® I 2000
Conditions
column CYCLOBOND I 2000, 25 cm x 4.6 mm I.D., 5 µm particles (20024AST)
column temp. 25 °C
mobile phase [A] 20 mM ammonium acetate, pH 4.1; [B] acetonitrile; (80:20, A:B)
flow rate 0.9 mL/min
sample 5 mg/mL in methanol
injection 10 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Chiral Applications, Phenols, Chemicals and Industrial Polymers, Oxygen compounds, Alcohols, Phenols & Diols
Featured Industry Chemicals and Industrial Polymers
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC