HPLC Analysis of Hydrobenzoin Enantiomers on Astec® CYCLOBOND® I 2000 RSP
HPLC Analysis of Hydrobenzoin Enantiomers on Astec® CYCLOBOND® I 2000 RSP
Conditions
column CYCLOBOND I 2000 RSP, 25 cm x 4.6 mm I.D., 5 µm particles (20324AST)
column temp. 25 °C
mobile phase [A] acetonitrile; [B] 10 mM ammonium acetate, pH 4.0; (25:75, A:B)
flow rate 1 mL/min
sample hydrobenzoin, 1 mg/mL in acetonitrile:water (50:50)
injection 5 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Alcohols
Featured Industry Chemicals and Industrial Polymers
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC