HPLC Analysis of Hydrolyzed β-Cyclodextrin on TSKgel® G-Oligo-PW
HPLC Analysis of Hydrolyzed β-Cyclodextrin on TSKgel® G-Oligo-PW
Conditions
column TSKgel G-Oligo-PW, 30 cm x 7.8 mm I.D., 6 µm particles (808031)
column temp. 60 °C
mobile phase distilled water
flow rate 1 mL/min
detector RI
Description
Categories Analytical Chromatography, Carbohydrates, Polysaccharides
Featured Industry Chemicals and Industrial Polymers
Food and Beverages
Legal Information TSKgel is a registered trademark of Tosoh Corporation