HPLC Analysis of α-Hydroxy Aliphatic Acids on Discovery® RP-Amide C16
HPLC Analysis of α-Hydroxy Aliphatic Acids on Discovery® RP-Amide C16
Conditions
column Discovery RP-AmideC16, 15 cm x 4.6 mm I.D., 5 µm particles (505013)
column temp. 30 C
mobile phase 25 mM potassium phosphate, pH 3.0
flow rate 1 ml/min
injection 10 µl
detector UV, 220 nm
Description
Categories Analytical Chromatography, Organic Acids
Featured Industry Chemicals and Industrial Polymers
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC